Alfa Aesar™ 7-Aminocephalosporanic acid, 98% (dry wt.), may cont. up to 2% water

Manufacturer: Alfa Aesar™ A1053003

Quantity 1g

 View more versions of this product

Catalog No. AAA1053003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 957-68-6
    Molecular Formula C10H12N2O5S
    Formula Weight 272.28
    MDL Number MFCD00005177
    Beilstein 622638
    Merck Index 14,434
    Synonym 7-ACA
    Chemical Name or Material 7-Aminocephalosporanic acid

    Specifications

    Formula Weight 272.28
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 957-68-6
    Beilstein 622638
    Merck Index 14,434
    Synonym 7-ACA
    Health Hazard 1 Danger
    Recommended Storage 2°C to 8°C
    Quantity 1g
    Concentration May contain up to 2% water
    Molecular Formula C10H12N2O5S
    Melting Point >300°C
    MDL Number MFCD00005177
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Chemical Name or Material 7-Aminocephalosporanic acid
    Assay 98%, (dry wt.)

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    2°C to 8°C

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit