Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminoheptanoic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
929-17-9
C7H15NO2
145.2
XDOLZJYETYVRKV-UHFFFAOYSA-N
7-aminoenanthic acid,heptanoic acid, 7-amino,omega-aminoenantic acid,aminoenanthic acid,amino-enanthylic acid,7-amino-heptanoic acid,7-aminohepentanoic acid,omega-aminoheptanoic acid,unii-b10hjx2t48,kyselina 7-aminoenanthova czech
13580
7-aminoheptanoic acid
C(CCCN)CCC(=O)O
929-17-9
C7H15NO2
145.2
XDOLZJYETYVRKV-UHFFFAOYSA-N
7-aminoenanthic acid,heptanoic acid, 7-amino,omega-aminoenantic acid,aminoenanthic acid,amino-enanthylic acid,7-amino-heptanoic acid,7-aminohepentanoic acid,omega-aminoheptanoic acid,unii-b10hjx2t48,kyselina 7-aminoenanthova czech
13580
7-aminoheptanoic acid
C(CCCN)CCC(=O)O
Packaging Glass Bottle
Infrared Spectrum Authentic
NMR Consistent with structure (1H−NMR)
Quantity 1g
Elemental Analysis 55.59 to 60.22% (C) 9.26 to 10.03% (N)
Melting Point 188.0°C to 189.5°C
Assay Percent Range 97%
Formula Weight 145.2
Percent Purity 97%
Chemical Name or Material 7-Aminoheptanoic acid

EINECSNumber : 213-197-5