Alfa Aesar™ (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%

Manufacturer: Alfa Aesar™ H5437803

Quantity 1g

 View more versions of this product

Catalog No. AAH5437803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 156311-83-0
    Molecular Formula C17H27F6N7OP2
    Formula Weight 521.38
    MDL Number MFCD03703417
    Synonym PyAOP
    Chemical Name or Material (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate

    Specifications

    Formula Weight 521.38
    Synonym PyAOP
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 163° to 168°C
    Quantity 1g
    Molecular Formula C17H27F6N7OP2
    MDL Number MFCD03703417
    CAS 156311-83-0
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number TH3890000
    Chemical Name or Material (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate
    Assay ≥99%

    Description

    Applications
    (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

    Solubility
    Soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit