Alfa Aesar™ 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole, 99%

Manufacturer: Alfa Aesar™ H5059203

Quantity 1g

 View more versions of this product

Catalog No. AAH5059203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 164736-47-4
    Molecular Formula C11H10BrN
    Formula Weight 236.11
    MDL Number MFCD00239009
    Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole

    Specifications

    Formula Weight 236.11
    Molecular Formula C11H10BrN
    MDL Number MFCD00239009
    CAS 164736-47-4
    Quantity 1g
    Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit