Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Bromo-1-heptanol, 96%

Manufacturer:  Alfa Aesar™ H5476203

Quantity 1g

 View more versions of this product

Catalog No. AAH5476203


  Check Availability
Add to cart

Specifications

Description

Applications
7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid. It can be used to prepare N-substituted [(carboxymethyl)phenoxy]ethyl benzoxazinones as PPARγ agonists and antidiabetic (type 2) agents. It can also be used to prepare thiochroman and chroman derivatives as pure antiestrogens.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Description & Specifications

Chemical Identifiers

10160-24-4
Br(CH2)7OH
MFCD00041708
C7H15BrO
195.1
7-Bromo-1-heptanol

Specifications

10160-24-4
1g
C7H15BrO
195.1
7-Bromo-1-heptanol
111° to 112°C (4mmHg)
113°C (235°F)
Br(CH2)7OH
MFCD00041708
96%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit