Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromoheptanoic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
30515-28-7
C7H13BrO2
209.08
JLPQXFFMVVPIRW-UHFFFAOYSA-N
7-bromo-heptanoic acid,heptanoic acid, 7-bromo,7-bromoenanthic acid,omega-bromoheptanoic acid,7-bromoheptanoicacid,5-bromopentylacetate;,7-bromo heptanoic acid,acmc-1aezo,ksc226m8n
121723
7-bromoheptanoic acid
C(CCCBr)CCC(=O)O
30515-28-7
C7H13BrO2
209.08
JLPQXFFMVVPIRW-UHFFFAOYSA-N
7-bromo-heptanoic acid,heptanoic acid, 7-bromo,7-bromoenanthic acid,omega-bromoheptanoic acid,7-bromoheptanoicacid,5-bromopentylacetate;,7-bromo heptanoic acid,acmc-1aezo,ksc226m8n
121723
7-bromoheptanoic acid
C(CCCBr)CCC(=O)O
Boiling Point 121.0°C to 125.0°C (5.0 mmHg)
Melting Point 29.0°C to 32.0°C
Quantity 25g
Infrared Spectrum Authentic
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material 7-Bromoheptanoic acid

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 250-225-5