Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Bromoquinoline, 95%

Click to view available options
Quantity:
250mg
1g
5g
4965-36-0
C9H6BrN
208.058
MFCD03695823
XYBSZCUHOLWQQU-UHFFFAOYSA-N
quinoline, 7-bromo,7-bromo-quinoline,pubchem12788,ksc238o9n,abbypharma ap-12-10883
521259
7-bromoquinoline
C1=CC2=C(C=C(C=C2)Br)N=C1
4965-36-0
C9H6BrN
208.058
MFCD03695823
XYBSZCUHOLWQQU-UHFFFAOYSA-N
quinoline, 7-bromo,7-bromo-quinoline,pubchem12788,ksc238o9n,abbypharma ap-12-10883
521259
7-bromoquinoline
C1=CC2=C(C=C(C=C2)Br)N=C1
Quantity 1g
Formula Weight 208.06
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 7-Bromoquinoline

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C