Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Chloro-4-hydroxyquinoline, 98+%

Click to view available options
Quantity:
25g
100g
86-99-7
C9H6ClNO
179.603
MFCD00006778
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol,7-chloro-4-hydroxyquinoline,7-chloro-4-quinolinol,7-chloroquinolin-4 1h-one,4-quinolinol, 7-chloro,4 1h-quinolinone, 7-chloro,7-chloro-quinolin-4-ol,7-chloro-4 1h-quinolinone,4mrw,7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
86-99-7
C9H6ClNO
179.603
MFCD00006778
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol,7-chloro-4-hydroxyquinoline,7-chloro-4-quinolinol,7-chloroquinolin-4 1h-one,4-quinolinol, 7-chloro,4 1h-quinolinone, 7-chloro,7-chloro-quinolin-4-ol,7-chloro-4 1h-quinolinone,4mrw,7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
Melting Point 278°C to 282°C
Quantity 25g
Beilstein 125356
Formula Weight 179.61
Percent Purity ≥98%
Chemical Name or Material 7-Chloro-4-hydroxyquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning