Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-4-hydroxyquinoline, 99%, Acros Organics

Click to view available options
Quantity:
25g
100g
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol,7-chloro-4-hydroxyquinoline,7-chloro-4-quinolinol,7-chloroquinolin-4 1h-one,4-quinolinol, 7-chloro,4 1h-quinolinone, 7-chloro,7-chloro-quinolin-4-ol,7-chloro-4 1h-quinolinone,4mrw,7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol,7-chloro-4-hydroxyquinoline,7-chloro-4-quinolinol,7-chloroquinolin-4 1h-one,4-quinolinol, 7-chloro,4 1h-quinolinone, 7-chloro,7-chloro-quinolin-4-ol,7-chloro-4 1h-quinolinone,4mrw,7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
Melting Point 276.0°C to 279.0°C
Quantity 100g
Packaging Glass bottle
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble
Formula Weight 179.61
Percent Purity 99%
Chemical Name or Material 7-Chloro-4-hydroxyquinoline