Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-4-hydroxyquinoline, 99%, Thermo Scientific™

Catalog Number p-4813282
Click to view available options
Quantity:
25 g
100 g
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol, 7-chloro-4-hydroxyquinoline, 7-chloro-4-quinolinol, 7-chloroquinolin-4 1h-one, 4-quinolinol, 7-chloro, 4 1h-quinolinone, 7-chloro, 7-chloro-quinolin-4-ol, 7-chloro-4 1h-quinolinone, 4mrw, 7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O
86-99-7
C9H6ClNO
179.61
XMFXTXKSWIDMER-UHFFFAOYSA-N
7-chloroquinolin-4-ol, 7-chloro-4-hydroxyquinoline, 7-chloro-4-quinolinol, 7-chloroquinolin-4 1h-one, 4-quinolinol, 7-chloro, 4 1h-quinolinone, 7-chloro, 7-chloro-quinolin-4-ol, 7-chloro-4 1h-quinolinone, 4mrw, 7-chloro4-quinolinol
66593
7-chloro-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)NC=CC2=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 276.0°C to 279.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 179.61
Percent Purity 99%
Chemical Name or Material 7-Chloro-4-hydroxyquinoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 201-715-2
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit