Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Chlorokynurenic acid sodium salt, 99+%

Click to view available options
Quantity:
10mg
50mg
1263094-00-3
C10H5ClNNaO3
245.594
MFCD12195840
IFZYIORLNGNLEI-UHFFFAOYSA-M
7-chlorokynurenic acid sodium salt
124219644
sodium;2-carboxy-7-chloroquinolin-4-olate
C1=CC2=C(C=C1Cl)N=C(C=C2[O-])C(=O)O.[Na+]
1263094-00-3
C10H5ClNNaO3
245.594
MFCD12195840
IFZYIORLNGNLEI-UHFFFAOYSA-M
7-chlorokynurenic acid sodium salt
124219644
sodium;2-carboxy-7-chloroquinolin-4-olate
C1=CC2=C(C=C1Cl)N=C(C=C2[O-])C(=O)O.[Na+]

NMDA receptor antagonist at the glycine site

Applications
7-Chlorokynurenic acid sodium salt is used as a Potent competitive inhibitor of L-glutamate transport into synaptic vesicles. It is also used as an NMDA receptor antagonist and inhibitor of EAAT.

Solubility
Soluble in water at 100mM

Notes
Store at room temperature (desiccating conditions). May be stored at 4°C. For long-term storage, aliquot and store at 4°C. Do not freeze. For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
Color White to Yellow
Quantity 50mg
Solubility Information Soluble in water at 100mM
Formula Weight 245.6
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material 7-Chlorokynurenic acid sodium salt