Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chlorokynurenic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

18000-24-3
C10H6ClNO3
223.612
UAWVRVFHMOSAPU-UHFFFAOYSA-N
7-Chloro-4-hydroxyquinoline-2-carboxylic acid
1884
CHEBI:63965

Potent competitive inhibitor of L-glutamate uptake

NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles. Also available as part of the NMDA Receptor - Glycine Site Tocriset™ . 7-Chlorokynurenic acid sodium salt (Cat. No. 3697) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
CAS 18000-24-3
Molecular Formula C10H6ClNO3
Synonym 7-Chloro-4-hydroxyquinoline-2-carboxylic acid
InChI Key UAWVRVFHMOSAPU-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1Cl)NC(=CC2=O)C(=O)O
IUPAC Name 7-chloro-4-oxo-1H-quinoline-2-carboxylic acid
Molecular Weight (g/mol) 223.612
PubChem CID 1884
ChEBI CHEBI:63965
Formula Weight 223.62
Chemical Name or Material 7-Chlorokynurenic acid
Grade >98%
Show More Show Less

Recommended Storage : Store at Room Temperature