Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloroquinoline, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
612-61-3
C9H6ClN
163.61
QNGUPQRODVPRDC-UHFFFAOYSA-N
quinoline, 7-chloro,7-chloro-quinoline,pubchem19899
521963
7-chloroquinoline
C1=CC2=C(C=C(C=C2)Cl)N=C1
612-61-3
C9H6ClN
163.61
QNGUPQRODVPRDC-UHFFFAOYSA-N
quinoline, 7-chloro,7-chloro-quinoline,pubchem19899
521963
7-chloroquinoline
C1=CC2=C(C=C(C=C2)Cl)N=C1
Quantity 5g
Assay Percent Range 97%
Formula Weight 163.61
Percent Purity 97%
Chemical Name or Material 7-Chloroquinoline

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning