Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Deazahypoxanthine, 97%

Click to view available options
Quantity:
1g
5g
25g
3680-71-5
C6H5N3O
135.126
MFCD08234923
FBMZEITWVNHWJW-UHFFFAOYSA-N
7h-pyrrolo 2,3-d pyrimidin-4-ol,pyrrolo 2,3-d pyrimidin-4-ol,7-deazahypoxanthine,3h-pyrrolo 2,3-d pyrimidin-4 7h-one,4-hydroxypyrrolo 2,3-d pyrimidine,4-hydroxypyrrolopyrimidine,1h-pyrrolo 2,3-d pyrimidin-4 7h-one,3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one,pyrrolo 2,3-d pyrimidin-4-one
96194
1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
C1=CNC2=C1C(=O)N=CN2
3680-71-5
C6H5N3O
135.126
MFCD08234923
FBMZEITWVNHWJW-UHFFFAOYSA-N
7h-pyrrolo 2,3-d pyrimidin-4-ol,pyrrolo 2,3-d pyrimidin-4-ol,7-deazahypoxanthine,3h-pyrrolo 2,3-d pyrimidin-4 7h-one,4-hydroxypyrrolo 2,3-d pyrimidine,4-hydroxypyrrolopyrimidine,1h-pyrrolo 2,3-d pyrimidin-4 7h-one,3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one,pyrrolo 2,3-d pyrimidin-4-one
96194
1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
C1=CNC2=C1C(=O)N=CN2
Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture and light sensitive. Store at -20°C
Sensitivity Light and moisture sensitive
Melting Point 345°C to 350°C
Quantity 1g
Color Brown
Beilstein 607869
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 135.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 7-Deazahypoxanthine

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P280i-P285-P304+P341-P305+P351+P338-P342+P311-P501c

H319-H334

RTECSNumber : UY9440000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen