Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Ethyl-10-hydroxycamptothecin, 98%, Acros Organics™

Click to view available options
Quantity:
100mg
1g
86639-52-3
C22H20N2O5
392.4
FJHBVJOVLFPMQE-QFIPXVFZSA-N
7-ethyl-10-hydroxycamptothecin,sn 38 lactone,7-ethyl-10-hydroxy-camptothecin,10-hydroxy-7-ethylcamptothecin,7-ethyl-10-hydroxycamptothecine,le-sn38,7-ethyl-10-hydroxy-20 s-camptothecin,captothecin, 7-ethyl-10-hydroxy,s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione,7-ethyl-10-hydroxy camptothecin
104842
CHEBI:8988
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O
86639-52-3
C22H20N2O5
392.4
FJHBVJOVLFPMQE-QFIPXVFZSA-N
7-ethyl-10-hydroxycamptothecin,sn 38 lactone,7-ethyl-10-hydroxy-camptothecin,10-hydroxy-7-ethylcamptothecin,7-ethyl-10-hydroxycamptothecine,le-sn38,7-ethyl-10-hydroxy-20 s-camptothecin,captothecin, 7-ethyl-10-hydroxy,s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione,7-ethyl-10-hydroxy camptothecin
104842
CHEBI:8988
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O
Quantity 100mg
Infrared Spectrum Authentic
Water 5% max.
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 392.4
Percent Purity 98%
Chemical Name or Material 7-Ethyl-10-hydroxycamptothecin

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Do not eat,drink or smoke when us

GHS Signal Word: Danger