Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, 96%

Click to view available options
Quantity:
250mg
1g
2018286-71-8
C14H17BFNO4
293.101
MIJNRXJMZCWQSY-UHFFFAOYSA-N
75487208
7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2F)OCC(=O)N3
2018286-71-8
C14H17BFNO4
293.101
MIJNRXJMZCWQSY-UHFFFAOYSA-N
75487208
7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2F)OCC(=O)N3
Quantity 1g
Formula Weight 293.1
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester