Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, 99%

Click to view available options
Quantity:
50mg
250mg
84806-27-9
C6H6FN3O4S
235.189
MFCD02682109
JXLHNMVSKXFWAO-UHFFFAOYSA-N
sbd-f,7-fluorobenzofurazan-4-sulfonic acid ammonium salt,ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate,ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate,azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate,ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate,7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane,2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
2737780
azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]
84806-27-9
C6H6FN3O4S
235.189
MFCD02682109
JXLHNMVSKXFWAO-UHFFFAOYSA-N
sbd-f,7-fluorobenzofurazan-4-sulfonic acid ammonium salt,ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate,ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate,azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate,ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate,7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane,2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
2737780
azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]
Quantity 250mg
Melting Point ~285°C to 295°C (decomposition)
Formula Weight 235.19
Percent Purity 99%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonic acid ammonium salt

Recommended Storage : 2° to 8°C