Alfa Aesar™ 7-Fluorochroman-4-one, may contain up to 15% 5-isomer, 98% (sum of isomers)

Manufacturer: Alfa Aesar™ H3188103

Quantity 1g

 View more versions of this product

Catalog No. AAH3188103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 113209-68-0
    Molecular Formula C9H7FO2
    Formula Weight 166.15
    MDL Number MFCD06202344
    Synonym 7-Fluoro-2,3-dihydrochromen-4-one
    Chemical Name or Material 7-Fluorochroman-4-one, sum of isomers

    Specifications

    Formula Weight 166.15
    Quantity 1g
    Molecular Formula C9H7FO2
    MDL Number MFCD06202344
    CAS 113209-68-0
    Synonym 7-Fluoro-2,3-dihydrochromen-4-one
    Melting Point 46° to 50°C
    Chemical Name or Material 7-Fluorochroman-4-one, sum of isomers
    Assay 98%, may contain up to 15% 5-isomer

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit