Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-(Fmoc-amino)heptanoic acid, 95%

Click to view available options
Quantity:
1g
5g
127582-76-7
C22H25NO4
367.445
MFCD01861353
FRPYXTFBPCYALT-UHFFFAOYSA-N
fmoc-7-ahp-oh,7-9h-fluoren-9-yl methoxy carbonyl amino heptanoic acid,7-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid,heptanoic acid, 7-9h-fluoren-9-ylmethoxy carbonyl amino,fmoc-ahp 7-oh,fmoc-nh-ch2 6-cooh,7-fmoc-amino enanthic acid,7-9h-fluoren-9-ylmethoxycarbonylamino heptanoic acid,fmoc-7-ahp-oh hplc,n-9-fluorenylmethoxycarbonyl-7-aminoenanthic acid
2761803
7-(9H-fluoren-9-ylmethoxycarbonylamino)heptanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCC(=O)O
127582-76-7
C22H25NO4
367.445
MFCD01861353
FRPYXTFBPCYALT-UHFFFAOYSA-N
fmoc-7-ahp-oh,7-9h-fluoren-9-yl methoxy carbonyl amino heptanoic acid,7-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid,heptanoic acid, 7-9h-fluoren-9-ylmethoxy carbonyl amino,fmoc-ahp 7-oh,fmoc-nh-ch2 6-cooh,7-fmoc-amino enanthic acid,7-9h-fluoren-9-ylmethoxycarbonylamino heptanoic acid,fmoc-7-ahp-oh hplc,n-9-fluorenylmethoxycarbonyl-7-aminoenanthic acid
2761803
7-(9H-fluoren-9-ylmethoxycarbonylamino)heptanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCCC(=O)O
Applications
7-(Fmoc-amino)heptanoic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
Quantity 5g
Color White
Linear Formula FmocNH(CH2)6CO2H
Formula Weight 367.44
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 7-(Fmoc-amino)heptanoic acid

storageNote1 : 2° to 8°C