Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Hexadecanone, 98+%

Click to view available options
Quantity:
5g
25g
100g
45206-91-5
C16H32O
240.431
MFCD00027354
XMIMKDVDMTZIIW-UHFFFAOYSA-N
7-hexadecanone,acmc-1as9q
98679
hexadecan-7-one
CCCCCCCCCC(=O)CCCCCC
45206-91-5
C16H32O
240.431
MFCD00027354
XMIMKDVDMTZIIW-UHFFFAOYSA-N
7-hexadecanone,acmc-1as9q
98679
hexadecan-7-one
CCCCCCCCCC(=O)CCCCCC
Melting Point 36°C to 38°C
Quantity 25g
Formula Weight 240.43
Percent Purity ≥98%
Chemical Name or Material 7-Hexadecanone