Alfa Aesar™ 7-Hexadecyn-1-ol, 95%

Manufacturer: Alfa Aesar™ B2211303

Quantity 1g

 View more versions of this product

Catalog No. AAB2211303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 822-21-9
    Molecular Formula C16H30O
    Linear Formula CH3(CH2)7C≡C(CH2)6OH
    Formula Weight 238.41
    MDL Number MFCD00041709
    Chemical Name or Material 7-Hexadecyn-1-ol

    Specifications

    Formula Weight 238.41
    Molecular Formula C16H30O
    Linear Formula CH3(CH2)7C≡C(CH2)6OH
    MDL Number MFCD00041709
    CAS 822-21-9
    Quantity 1g
    Chemical Name or Material 7-Hexadecyn-1-ol
    Assay 95%

    Description

    Applications
    7-Hexadecyn-1-ol is used in the preparation of 15-hexadecynoic acid. It acts as a reagent for pharmaceutical research and in chemical synthesis.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit