Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-1,2,3,4-tetrahydroquinoline, 96%, Acros Organics™

Click to view available options
Quantity:
1g
5g
58196-33-1
C9H11NO
149.19
HJJRGZMJZDSMDB-UHFFFAOYSA-N
7-hydroxy-1,2,3,4-tetrahydroquinoline,7-quinolinol, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydro-7-quinolinol,1,2,3,4-tetrahydro-quinolin-7-ol,1,2,3,4-tetrahydrochinolin-7-ol,pubchem12787,7-hydroxy-3,4-dihydro-2h-quinoline,1,2,3,4-tetrahydro-7-hydroxyquinoline,7-hydroxy-1,2,3,4-tetrahydro-quinolin,7-hydroxy-1,2,3,4-tetrahydro quinolinoe
93980
1,2,3,4-tetrahydroquinolin-7-ol
C1CC2=C(C=C(C=C2)O)NC1
58196-33-1
C9H11NO
149.19
HJJRGZMJZDSMDB-UHFFFAOYSA-N
7-hydroxy-1,2,3,4-tetrahydroquinoline,7-quinolinol, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydro-7-quinolinol,1,2,3,4-tetrahydro-quinolin-7-ol,1,2,3,4-tetrahydrochinolin-7-ol,pubchem12787,7-hydroxy-3,4-dihydro-2h-quinoline,1,2,3,4-tetrahydro-7-hydroxyquinoline,7-hydroxy-1,2,3,4-tetrahydro-quinolin,7-hydroxy-1,2,3,4-tetrahydro quinolinoe
93980
1,2,3,4-tetrahydroquinolin-7-ol
C1CC2=C(C=C(C=C2)O)NC1
Infrared Spectrum Authentic
Melting Point 91.0°C to 93.5°C
Quantity 5g
Assay Percent Range 95% min. (HPLC)
Formula Weight 149.19
Percent Purity 96%
Chemical Name or Material 7-Hydroxy-1,2,3,4-tetrahydroquinoline

GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning