Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-4-cholesten-3-one, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

7 α-Hydroxy-4-cholesten-3-one (C4) is known as a marker for the cholesterol 7 α-hydroxylase activity. It plays an important role in bile acid metabolism.

Specifications

Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Linear Formula C27H44O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4101
View Documents
MilliporeSigma Supelco
528531MG
1 mg N/A
11-101-4102
View Documents
MilliporeSigma Supelco
528535MG
5 mg N/A
Description

Description

Specifications

Specifications

≥95.0% (HPLC)
Analytical Standard
C27H44O2
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit