Learn More
Aat BioquestSupplier Diversity Partner 7-Hydroxy-4-methylcoumarin-3-acetic acid *CAS 5852-10-8* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 554
7-Hydroxy-4-methylcoumarin-3-acetic acid is a blue fluorophore that has pH-dependent and environment-sensitive fluorescence. This coumarin is increasingly used to label peptides, nucleotides and carbohydrates.
Catalog No. 50-195-2927