Learn More
Aat BioquestSupplier Diversity Partner 7-Hydroxycoumarin-3-carboxylic acid *CAS 779-27-1* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 550
7-Hydroxycoumarin-3-carboxylic acid is a blue fluorophores for labeling proteins and nucleic acids mostly through the in situ formation of its succinimidyl ester catalyzed by EDAC.
Catalog No. 50-195-2923