Learn More
Aat BioquestSupplier Diversity Partner 7-Hydroxycoumarin-3-carboxylic acid, succinimidyl ester *CAS 134471-24-2* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 551
7-Hydroxycoumarin-3-carboxylic acid, SE is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It is a blue fluorescent dyes for labeling proteins and nucleic acids.
Catalog No. 50-195-2924