Learn More
Aat BioquestSupplier Diversity Partner 7-Hydroxycoumarin-4-acetic acid, succinimidyl ester DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 553
7-Hydroxycoumarin-4-acetic acid is used for labeling peptides and proteins to give blue fluorescence. It can also be used for labeling other biological molecules.
Catalog No. 50-195-2926