Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxyflavone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
6665-86-7
C15H10O3
238.242
MFCD00006835
MQGPSCMMNJKMHQ-UHFFFAOYSA-N
7-hydroxyflavone,flavone, 7-hydroxy,7-hydroxy-2-phenyl-4-benzopyrone,4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl,7-hydroxy-2-phenyl-4h-chromen-4-one,4h-1-benzopyran-4-one, 7-hydroxy,unii-ze72458e4l,7-hydroxy-2-phenyl-chromen-4-one,7-hydroxy-2-phenyl-4h-1-benzopyran-4-one,7-hydroxyflavone, 11
5281894
CHEBI:2268
7-hydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O
6665-86-7
C15H10O3
238.242
MFCD00006835
MQGPSCMMNJKMHQ-UHFFFAOYSA-N
7-hydroxyflavone,flavone, 7-hydroxy,7-hydroxy-2-phenyl-4-benzopyrone,4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl,7-hydroxy-2-phenyl-4h-chromen-4-one,4h-1-benzopyran-4-one, 7-hydroxy,unii-ze72458e4l,7-hydroxy-2-phenyl-chromen-4-one,7-hydroxy-2-phenyl-4h-1-benzopyran-4-one,7-hydroxyflavone, 11
5281894
CHEBI:2268
7-hydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O
Color White
Infrared Spectrum Authentic
Quantity 5g
Melting Point 243°C to 247°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 238.24
Physical Form Crystalline
Percent Purity ≥97.5%
Chemical Name or Material 7-Hydroxyflavone, 98%
Show More Show Less

EINECSNumber : 229-705-3