Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxyisoflavone 98.0+%, TCI America™

missing translation for 'orderingAttributeHoverText'
Quantity:
1g
5g
13057-72-2
C15H10O3
238.242
MFCD00017701
WMKOZARWBMFKAS-UHFFFAOYSA-N
7-Hydroxy-3-phenylchromone
5376891
CHEBI:12256
7-hydroxy-3-phenylchromen-4-one
C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O
13057-72-2
C15H10O3
238.242
MFCD00017701
WMKOZARWBMFKAS-UHFFFAOYSA-N
7-Hydroxy-3-phenylchromone
5376891
CHEBI:12256
7-hydroxy-3-phenylchromen-4-one
C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O
Color White-Yellow
Melting Point 216°C
Quantity 5g
Formula Weight 238.24
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 7-Hydroxyisoflavone

RTECSNumber : DJ3100255

TSCA : No