Alfa Aesar™ 7-Iodoisatin, 97%

Manufacturer: Alfa Aesar™ H5234403

Quantity 1g

 View more versions of this product

Catalog No. AAH5234403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20780-78-3
    Molecular Formula C8H4INO2
    Formula Weight 273.03
    MDL Number MFCD01248407
    Synonym 7-Iodo-1H-indole-2,3-dione
    Chemical Name or Material 7-Iodoisatin

    Specifications

    Formula Weight 273.03
    Molecular Formula C8H4INO2
    MDL Number MFCD01248407
    CAS 20780-78-3
    Quantity 1g
    Synonym 7-Iodo-1H-indole-2,3-dione
    Chemical Name or Material 7-Iodoisatin
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit