Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Iodooxindole, 96%

Manufacturer:  Alfa Aesar™ H5233703

Quantity 1g

 View more versions of this product

Catalog No. AAH5233703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

31676-49-0
259.04
7-Iodo-2-indolinone
C8H6INO
MFCD09835634
7-Iodooxindole

Specifications

31676-49-0
259.04
208° to 209°C
1g
96%
C8H6INO
MFCD09835634
7-Iodo-2-indolinone
7-Iodooxindole
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit