Learn More
7-Ketocholesterol Mouse anti-Human, ATTO 655, Clone: 7E1, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB19298
Description
7-Ketocholesterol Monoclonal antibody specifically detects 7-Ketocholesterol in Human samples. It is validated for WB, ELISA, ICC, IF.Specifications
7-Ketocholesterol | |
7E1 | |
Liquid | |
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary | |
Human |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
ATTO 655 | |
Mouse | |
IgG2b | |
Store at -20°C | |
Monoclonal |