Learn More
7-Ketocholesterol Mouse anti-Non-species specific, Biotin, Clone: 3F7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259357B
Catalog No. NBP259357B
Add to cart
Description
Description
7-Ketocholesterol Monoclonal antibody specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
7-Ketocholesterol
3F7
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG3
100 ul
Primary
Specific for 7-ketocholesterol modified proteins.