Learn More
7-Ketocholesterol Antibody (7E1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259356UV
Description
7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
7-Ketocholesterol | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
70 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
7KC, 7-KC, 7KC-Modified KLH | |
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for 7-ketocholesterol modified proteins. | |
Store at 4C in the dark. | |
IgG2b |