Learn More
7-Ketocholesterol Mouse anti-Non-species specific, DyLight 550, Clone: 70, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259356R
Catalog No. NBP259356R
Add to cart
Description
Description
7-Ketocholesterol Monoclonal antibody specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
7-Ketocholesterol
70
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2b
100 ul
Primary
Specific for 7-ketocholesterol modified proteins.