Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%

Click to view available options
Quantity:
250mg
1g
5g
1810-74-8
C13H17NO
203.285
MFCD00627122
VNIQAUZZZWOJPT-UHFFFAOYSA-N
7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline,2,2,4-trimethyl-7-methoxy-1,2-dihydroquinoline,quinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethyl,1,2-dihydro-2,2,4-trimethyl-7-methoxyquinoline,1,2-dihydro-7-methoxy-2,2,4-trimethylquinoline,7-methoxy-2,2,4-trimethyl-1,2-dihydro-quinoline,quinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro
643478
7-methoxy-2,2,4-trimethyl-1H-quinoline
CC1=CC(NC2=C1C=CC(=C2)OC)(C)C
1810-74-8
C13H17NO
203.285
MFCD00627122
VNIQAUZZZWOJPT-UHFFFAOYSA-N
7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline,2,2,4-trimethyl-7-methoxy-1,2-dihydroquinoline,quinoline, 1,2-dihydro-7-methoxy-2,2,4-trimethyl,1,2-dihydro-2,2,4-trimethyl-7-methoxyquinoline,1,2-dihydro-7-methoxy-2,2,4-trimethylquinoline,7-methoxy-2,2,4-trimethyl-1,2-dihydro-quinoline,quinoline, 7-methoxy-2,2,4-trimethyl-1,2-dihydro
643478
7-methoxy-2,2,4-trimethyl-1H-quinoline
CC1=CC(NC2=C1C=CC(=C2)OC)(C)C
Quantity 1g
Beilstein 1426616
Formula Weight 203.29
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline