Alfa Aesar™ 7-Methoxy-2-tetralone, 98%

Manufacturer: Alfa Aesar™ B22432-03

Quantity 1g

 View more versions of this product

Catalog No. AAB2243203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4133-34-0
    Molecular Formula C11H12O2
    Formula Weight 176.22g/mol
    MDL Number MFCD00001730
    Chemical Name or Material 7-Methoxy-2-tetralone

    Specifications

    Boiling Point 124° to 126°C (1.5mmHg)
    Quantity 1g
    Flash Point >110°C (230°F)
    CAS 4133-34-0
    Molecular Formula C11H12O2
    Formula Weight 176.22g/mol
    MDL Number MFCD00001730
    Recommended Storage 2° to 8°C
    Chemical Name or Material 7-Methoxy-2-tetralone
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit