Alfa Aesar™ 7-Methoxy-4-methyl-2(1H)-quinolinone, 96%

Manufacturer: Alfa Aesar™ H3167603

Quantity 1g

 View more versions of this product

Catalog No. AAH3167603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 40053-37-0
    Molecular Formula C11H11NO2
    Formula Weight 189.22
    MDL Number MFCD00711756
    Chemical Name or Material 7-Methoxy-4-methyl-2(1H)-quinolinone

    Specifications

    Formula Weight 189.22
    Molecular Formula C11H11NO2
    MDL Number MFCD00711756
    CAS 40053-37-0
    Melting Point 205° to 207°C
    Quantity 1g
    Chemical Name or Material 7-Methoxy-4-methyl-2(1H)-quinolinone
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit