Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Methoxy-4-methyl-2(1H)-quinolinone, 96%

Click to view available options
Quantity:
250mg
1g
40053-37-0
C11H11NO2
189.214
MFCD00711756
MXLWDNBRJGOMLA-UHFFFAOYSA-N
7-methoxy-4-methyl-quinolin-2-ol,7-methoxy-4-methylquinolin-2 1h-one,7-methoxy-4-methyl-2 1h-quinolinone,7-methoxy-4-methylquinolin-2-ol,enamine_005460,cambridge id 5511456,7-methoxy-4-methyl-2-hydroxyquinoline,7-methoxy-4-methylhydroquinolin-2-one,7-methoxy-4-methylquinolin-2 1h one,2-hydroxy-7-methoxy-4-methylquinoline
761548
7-methoxy-4-methyl-1H-quinolin-2-one
CC1=CC(=O)NC2=C1C=CC(=C2)OC
40053-37-0
C11H11NO2
189.214
MFCD00711756
MXLWDNBRJGOMLA-UHFFFAOYSA-N
7-methoxy-4-methyl-quinolin-2-ol,7-methoxy-4-methylquinolin-2 1h-one,7-methoxy-4-methyl-2 1h-quinolinone,7-methoxy-4-methylquinolin-2-ol,enamine_005460,cambridge id 5511456,7-methoxy-4-methyl-2-hydroxyquinoline,7-methoxy-4-methylhydroquinolin-2-one,7-methoxy-4-methylquinolin-2 1h one,2-hydroxy-7-methoxy-4-methylquinoline
761548
7-methoxy-4-methyl-1H-quinolin-2-one
CC1=CC(=O)NC2=C1C=CC(=C2)OC
Quantity 250mg
Melting Point 205°C to 207°C
Formula Weight 189.22
Percent Purity 96%
Chemical Name or Material 7-Methoxy-4-methyl-2(1H)-quinolinone