Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-4-methylcoumarin, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
2555-28-4
C11H10O3
190.198
MFCD00009773
UDFPKNSWSYBIHO-UHFFFAOYSA-N
7-methoxy-4-methylcoumarin,7-methoxy-4-methyl-2h-chromen-2-one,2h-1-benzopyran-2-one, 7-methoxy-4-methyl,4-methylherniarin,hymecromone methyl ether,herniarin, 4-methyl,7-methoxy-4-methyl-chromen-2-one,4-methyl-7-methoxycoumarin,coumarin,4-methyl-7-methoxy,coumarin, 7-methoxy-4-methyl
390807
7-methoxy-4-methylchromen-2-one
CC1=CC(=O)OC2=C1C=CC(=C2)OC
2555-28-4
C11H10O3
190.198
MFCD00009773
UDFPKNSWSYBIHO-UHFFFAOYSA-N
7-methoxy-4-methylcoumarin,7-methoxy-4-methyl-2h-chromen-2-one,2h-1-benzopyran-2-one, 7-methoxy-4-methyl,4-methylherniarin,hymecromone methyl ether,herniarin, 4-methyl,7-methoxy-4-methyl-chromen-2-one,4-methyl-7-methoxycoumarin,coumarin,4-methyl-7-methoxy,coumarin, 7-methoxy-4-methyl
390807
7-methoxy-4-methylchromen-2-one
CC1=CC(=O)OC2=C1C=CC(=C2)OC
Color White to Yellow
Packaging Glass bottle
Quantity 1g
Melting Point 157.0°C to 161.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 190.2
Physical Form Powder, Crystals, Crystalline Powder and/or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 7-Methoxy-4-methylcoumarin
Show More Show Less