Alfa Aesar™ 7-Methoxy-4-methylcoumarin, 99%

Manufacturer: Alfa Aesar™ A1178506

Quantity 5g

 View more versions of this product

Catalog No. AAA1178506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2555-28-4
    Molecular Formula C11H10O3
    Formula Weight 190.2
    MDL Number MFCD00009773
    Beilstein 156900
    Synonym Methyl 4-methylumbelliferyl ether
    Chemical Name or Material 7-Methoxy-4-methylcoumarin

    Specifications

    Formula Weight 190.2
    MDL Number MFCD00009773
    Beilstein 156900
    Molecular Formula C11H10O3
    CAS 2555-28-4
    Synonym Methyl 4-methylumbelliferyl ether
    Melting Point 159° to 162°C
    Quantity 5g
    Chemical Name or Material 7-Methoxy-4-methylcoumarin
    Assay 99%

    Description

    Applications
    A coumarin derivative and fluorescent label.

    Solubility
    Soluble in Methanol. Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit