Alfa Aesar™ 7-Methoxy-4-methylquinoline, 98%

Manufacturer: Alfa Aesar™ H32486ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH32486ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6238-12-6
    Molecular Formula C11H11NO
    Formula Weight 173.21
    MDL Number MFCD11655579
    Chemical Name or Material 7-Methoxy-4-methylquinoline

    Specifications

    Formula Weight 173.21
    Molecular Formula C11H11NO
    MDL Number MFCD11655579
    CAS 6238-12-6
    Quantity 500mg
    Chemical Name or Material 7-Methoxy-4-methylquinoline
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit