Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxybenzofuran-2-carboxylic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4790-79-8
C10H8O4
192.17
MFCD00060516
UOCNTRAAJNWDND-UHFFFAOYSA-N
7-methoxybenzofuran-2-carboxylic acid,7-methoxybenzo b furan-2-carboxylic acid,2-benzofurancarboxylic acid, 7-methoxy,7-methoxy-2-benzofurancarboxylic acid,7-methoxy-benzofuran-2-carboxylic acid,7-methoxybenzofuran-2-carboxylicacid,acmc-1am7f,ksc236k1t,7methoxybenzofuran-2-carboxylic acid
606037
7-methoxy-1-benzofuran-2-carboxylic acid
COC1=CC=CC2=C1OC(=C2)C(=O)O
4790-79-8
C10H8O4
192.17
MFCD00060516
UOCNTRAAJNWDND-UHFFFAOYSA-N
7-methoxybenzofuran-2-carboxylic acid,7-methoxybenzo b furan-2-carboxylic acid,2-benzofurancarboxylic acid, 7-methoxy,7-methoxy-2-benzofurancarboxylic acid,7-methoxy-benzofuran-2-carboxylic acid,7-methoxybenzofuran-2-carboxylicacid,acmc-1am7f,ksc236k1t,7methoxybenzofuran-2-carboxylic acid
606037
7-methoxy-1-benzofuran-2-carboxylic acid
COC1=CC=CC2=C1OC(=C2)C(=O)O
Infrared Spectrum Authentic
Quantity 1g
Melting Point 215°C to 220°C
Color Beige
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 192.17
Physical Form Fine Crystals
Percent Purity ≥96.0%
Chemical Name or Material 7-Methoxybenzofuran-2-carboxylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.