Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Methoxyflavanone, 98%

Click to view available options
Quantity:
1g
5g
21785-09-1
C16H14O3
254.285
MFCD00017486
VYESEQLQFXUROZ-UHFFFAOYSA-N
7-methoxyflavanone,7-methoxy-2-phenylchroman-4-one,chembl9687,7-methoxy-2-phenyl-chroman-4-one,7-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one,acmc-1cngn,maybridge3_004341,+/--7-methoxy-2-phenylchroman-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-7-methoxy-2-phenyl
2825024
7-methoxy-2-phenyl-2,3-dihydrochromen-4-one
COC1=CC2=C(C=C1)C(=O)CC(O2)C3=CC=CC=C3
21785-09-1
C16H14O3
254.285
MFCD00017486
VYESEQLQFXUROZ-UHFFFAOYSA-N
7-methoxyflavanone,7-methoxy-2-phenylchroman-4-one,chembl9687,7-methoxy-2-phenyl-chroman-4-one,7-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one,acmc-1cngn,maybridge3_004341,+/--7-methoxy-2-phenylchroman-4-one,4h-1-benzopyran-4-one, 2,3-dihydro-7-methoxy-2-phenyl
2825024
7-methoxy-2-phenyl-2,3-dihydrochromen-4-one
COC1=CC2=C(C=C1)C(=O)CC(O2)C3=CC=CC=C3
Quantity 5g
Melting Point 86°C to 90°C
Formula Weight 254.29
Percent Purity 98%
Chemical Name or Material 7-Methoxyflavanone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning