Alfa Aesar™ 7-Methoxyflavone, 99%

Manufacturer: Alfa Aesar™ B2485006

Quantity 5g

 View more versions of this product

Catalog No. AAB2485006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22395-22-8
    Molecular Formula C16H12O3
    Formula Weight 252.27
    MDL Number MFCD00017462
    Chemical Name or Material 7-Methoxyflavone

    Specifications

    Formula Weight 252.27
    Molecular Formula C16H12O3
    MDL Number MFCD00017462
    CAS 22395-22-8
    Melting Point 108° to 112°C
    Quantity 5g
    Chemical Name or Material 7-Methoxyflavone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit