Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 7-Methoxyflavone, 99%

Manufacturer:  Alfa Aesar™ B2485014

Quantity 25g

 View more versions of this product

Catalog No. AAB2485014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

22395-22-8
252.27
7-Methoxyflavone
C16H12O3
MFCD00017462

Specifications

108° to 112°C
22395-22-8
252.27
7-Methoxyflavone
25g
C16H12O3
MFCD00017462
99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit