Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine-4-thiol, 96%, Thermo Scientific™

Click to view available options
Quantity:
500mg
314260-78-1
C11H12N2S2
236.351
MFCD02679296
RBWNDBNSJFCLBZ-UHFFFAOYSA-N
7-methyl-5,6,7,8-tetrahydro-benzo 4,5 thieno 2,3-d pyrimidine-4-thiol, 7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine-4 3h-thione, 7-methyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine-4-thiol, 1 benzothieno 2,3-d pyrimidine-4 3h-thione,5,6,7,8-tetrahydro-7-methyl, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,5-triene-3-thione, 7-methyl-3,5,6,7,8-pentahydrobenzo b thiopheno 2,3-d pyrimidine-4-thione, 7-methyl-5,6,7,8-tetrahydro-3h-1 benzothiolo 2,3-d pyrimidine-4-thione, 7-methyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine-4-thiol, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene-3-thiol, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0?,? trideca-1 9 ,2,4,6-tetraene-3-thiol
2790355
7-methyl-5,6,7,8-tetrahydro-3H-[1]benzothiolo[2,3-d]pyrimidine-4-thione
CC1CCC2=C(C1)SC3=C2C(=S)NC=N3
314260-78-1
C11H12N2S2
236.351
MFCD02679296
RBWNDBNSJFCLBZ-UHFFFAOYSA-N
7-methyl-5,6,7,8-tetrahydro-benzo 4,5 thieno 2,3-d pyrimidine-4-thiol, 7-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine-4 3h-thione, 7-methyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine-4-thiol, 1 benzothieno 2,3-d pyrimidine-4 3h-thione,5,6,7,8-tetrahydro-7-methyl, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,5-triene-3-thione, 7-methyl-3,5,6,7,8-pentahydrobenzo b thiopheno 2,3-d pyrimidine-4-thione, 7-methyl-5,6,7,8-tetrahydro-3h-1 benzothiolo 2,3-d pyrimidine-4-thione, 7-methyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine-4-thiol, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 , trideca-1 9 ,2,4,6-tetraene-3-thiol, 11-methyl-8-thia-4,6-diazatricyclo 7.4.0.0?,? trideca-1 9 ,2,4,6-tetraene-3-thiol
2790355
7-methyl-5,6,7,8-tetrahydro-3H-[1]benzothiolo[2,3-d]pyrimidine-4-thione
CC1CCC2=C(C1)SC3=C2C(=S)NC=N3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 220°C to 225°C
Quantity 500mg
Formula Weight 236.36
Percent Purity 96%
Chemical Name or Material 7-Methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine-4-thiol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures