Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methylindole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
933-67-5
C9H9N
131.178
KGWPHCDTOLQQEP-UHFFFAOYSA-N
7-methylindole,7-methylindol,1h-indole, 7-methyl,indole, 7-methyl,7-methyindole,7-methyl-indole,7-methyl indole,unii-7z1e6hit9s,7z1e6hit9s,7-methyl-1h-indol-1-yl
70275
7-methyl-1H-indole
CC1=CC=CC2=C1NC=C2
933-67-5
C9H9N
131.178
KGWPHCDTOLQQEP-UHFFFAOYSA-N
7-methylindole,7-methylindol,1h-indole, 7-methyl,indole, 7-methyl,7-methyindole,7-methyl-indole,7-methyl indole,unii-7z1e6hit9s,7z1e6hit9s,7-methyl-1h-indol-1-yl
70275
7-methyl-1H-indole
CC1=CC=CC2=C1NC=C2
Color Beige to White
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 80°C to 83°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, II, 205
Formula Weight 131.18
Physical Form Shiny Flakes
Percent Purity ≥98.5%
Chemical Name or Material 7-Methylindole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning