Alfa Aesar™ 7-Methylxanthine, 98%

Manufacturer: Alfa Aesar™ H2919003

Quantity 1g

 View more versions of this product

Catalog No. AAH2919003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 552-62-5
    Molecular Formula C6H6N4O2
    Formula Weight 166.14
    MDL Number MFCD00037979
    Synonym 2,6-Dihydroxy-7-methylpurine; Heteroxanthine
    Chemical Name or Material 7-Methylxanthine

    Specifications

    Formula Weight 166.14
    Molecular Formula C6H6N4O2
    MDL Number MFCD00037979
    CAS 552-62-5
    Synonym 2,6-Dihydroxy-7-methylpurine; Heteroxanthine
    Melting Point >300°C
    Quantity 1g
    Chemical Name or Material 7-Methylxanthine
    Assay 98%

    Description

    Applications
    It is used to metabolite theophylline and caffeine.

    Solubility
    Soluble in dimethyl sulfoxide. Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit