Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-NINA, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

161467-34-1
C7H4N3NaO2
185.118
DJMLKKIIBMJGIQ-UHFFFAOYSA-N
7-Nitroindazole monosodium salt
23661758
sodium;7-nitroindazol-2-ide
C1=CC2=C[N-]N=C2C(=C1)[N+](=O)[O-].[Na+]

Non-selective NOS inhibitor. Sodium salt of 7-nitroindazole (Cat. No. 0602)

Sodium salt of the non-selective NOS inhibitor 7-nitroindazole (Cat. No. 0602).
Quantity 50mg
Formula Weight 185.12
Chemical Name or Material 7-NINA
Grade >98%

Recommended Storage : Desiccate at -20°C